4-Cyanobenzenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14081-14
A14081-03
CAAAAA14081-14EA
1233.98
CAD
CAAAAA14081-14
CAAAA14081-03
4-Cyanobenzenesulfonyl chloride 97%
4-Cyanobenzenesulphonyl chloride
Purity: 97%
CAS No.: 49584-26-1
Molecular Formula: C7H4ClNO2S
Formula Weight: 201.63
Melting Point: 107-110°
Beilstein Reference: 777939
UN No.: 3261
MDL No.: MFCD00052337
Formula:
C₇H₄ClNO₂S Melting Pt: 107…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052337 CAS Number: 49584-26-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...