2,3-Dimethoxybenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14080-14 A14080-22
CAAAA14080-14EA 71.89 CAD
CAAAA14080-14 CAAAA14080-22 CAAAA14080-36
2,3-Dimethoxybenzaldehyde 98+%
2,3-Dimethoxybenzaldehyde

Purity: 98+% CAS No.: 86-51-1 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 51-52° Boiling Point: 178°/20mm Beilstein Reference: 908264 MDL No.: MFCD00003309

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 178 °C/20 mm
Melting Pt: 51…52 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003309
CAS Number: 86-51-1
EINECS: 201-677-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR