3-Chlorobenzylamine 97+%

Supplier: Thermo Scientific Chemicals
Danger

A14078-09 A14078-18
CAAAA14078-09EA 129.56 CAD
CAAAA14078-09 CAAAA14078-18
3-Chlorobenzylamine 97+%
3-Chlorobenzylamine

Purity: 97+% CAS No.: 4152-90-3 Molecular Formula: C7H8ClN Formula Weight: 141.60 Flash Point: 98°(208°F) Refractive Index: 1.5590 Odor: Amine-like Boiling Point: 226° Beilstein Reference: 774509 UN No.: 2735 MDL No.: MFCD00040752

Formula: C₇H₈ClN
Boiling Pt: 226 °C
Density: 1.159
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040752
CAS Number: 4152-90-3
EINECS: 223-985-0
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR