4-tert-Butylbenzonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14076-03 A14076-06 A14076-14
CAAAA14076-03EA 59.25 CAD
CAAAA14076-03 CAAAA14076-06 CAAAA14076-14
4-tert-Butylbenzonitrile 98+%
4-tert-Butylbenzonitrile

Purity: 98+% CAS No.: 4210-32-6 Molecular Formula: C11H13N Formula Weight: 159.23 Flash Point: 110°(230°F) Refractive Index: 1.5170 Odor: Characteristic Boiling Point: 258° Beilstein Reference: 2080049 MDL No.: MFCD00075840

Formula: C₁₁H₁₃N
Boiling Pt: 258 °C
Density: 0.940
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075840
CAS Number: 4210-32-6
EINECS: 224-137-2
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR