3,5-Diaminobenzoic acid 98% may contain up to 3% moisture
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14075-22
A14075-36
A14075-18
CAAAAA14075-22EA
86.43
CAD
CAAAAA14075-22
CAAAAA14075-36
CAAAA14075-18
3,5-Diaminobenzoic acid 98% may contain up to 3% moisture
3,5-Diaminobenzoic acid
Purity: 98%
CAS No.: 535-87-5
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Flash Point: 210°(410°F)
Melting Point: ca 235° dec.
Beilstein Reference: 2086484
Merck Reference: 14,2978
MDL No.: MFCD00007807
Formula:
C₇H₈N₂O₂ Boiling Pt: 546 °C (1013 hPa) Melting Pt: ca 235 °C (dec.) Density: 1.45 g/cm³ (20 °C) Flash Pt: 210 °C(410 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007807 CAS Number: 535-87-5 EINECS: 208-621-0 Merck Index: 14,2978 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...