5-Chlorosalicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-2-hydroxybenzaldehyde

A14069-22 A14069-06 A14069-14
CAAAAA14069-22EA 530.88 CAD
CAAAAA14069-22 CAAAA14069-06 CAAAA14069-14
5-Chlorosalicylaldehyde 98%
5-Chlorosalicylaldehyde

Purity: 98% CAS No.: 635-93-8 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Melting Point: 99-103° Beilstein Reference: 636632

Formula: C₇H₅ClO₂
Boiling Pt: 105 °C (12 -rr)
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00003331
CAS Number: 635-93-8
EINECS: 211-244-4
UN: 3077
ADR: "9",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR