4-Hydroxybenzylacetone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Raspberry ketone, 1-(4-Hydroxyphenyl)-3-butanone, 4-(4-Hydroxyphenyl)-2-butanone
A14064-30
A14064-14
A14064-22
CAAAAA14064-30EA
451.88
CAD
CAAAAA14064-30
CAAAA14064-14
CAAAA14064-22
4-Hydroxybenzylacetone 98%
4-Hydroxybenzylacetone
Purity: 98%
CAS No.: 5471-51-2
Molecular Formula: C10H12O2
Formula Weight: 164.20
Melting Point: 80-84°
Beilstein Reference: 776080
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 292 Melting Pt: 80…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002394 CAS Number: 5471-51-2 EINECS: 226-806-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...