Benzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14062-0B
A14062-0I
A14062-30
CAAAA14062-0BEA
79.79
CAD
CAAAA14062-0B
CAAAA14062-0I
CAAAA14062-30
Benzoic acid 99%
Benzoic acid
Purity: 99%
CAS No.: 65-85-0
Molecular Formula: C7H6O2
Formula Weight: 122.12
Flash Point: 121°(250°F)
Physical Form: Crystalline powder
Density: 1.32
Appearance: White
Odor: Weak, characteristic
pH: 2.5-3.5
Melting Point: 121-124°
Boiling Point: 249°
Beilstein Reference: 636131
Merck Reference: 14,1091
MDL No.: MFCD00002398
Formula:
C₇H₆O₂ Boiling Pt: 249 °C Melting Pt: 121…124 °C Density: 1.32 Flash Pt: 121 °C(250 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 EINECS: 200-618-2 REACH: 01-2119455536-33 Merck Index: 14,1091 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...