2-Chlorobenzenesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14054-06
A14054-14
CAAAA14054-06EA
124.66
CAD
CAAAA14054-06
CAAAA14054-14
CAAAA14054-22
2-Chlorobenzenesulfonyl chloride 98%
2-Chlorobenzenesulphonyl chloride
Purity: 98%
CAS No.: 2905-23-9
Molecular Formula: C6H4Cl2O2S
Formula Weight: 211.07
Flash Point: 140°(284°F)
Refractive Index: 1.5760
Melting Point: 26-28°
Boiling Point: 150-152°/12mm
Beilstein Reference: 974321
UN No.: 3265
MDL No.: MFCD00051520
Formula:
C₆H₄Cl₂O₂S Boiling Pt: 150…152 °C/12 mm Melting Pt: 26…28 °C Density: 1.548 Flash Pt: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051520 CAS Number: 2905-23-9 EINECS: 220-807-3 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...