1-Bromo-3,5-difluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14053-30
A14053-06
A14053-14
A14053-22
CAAAAA14053-30EA
1300.34
CAD
CAAAAA14053-30
CAAAA14053-06
CAAAA14053-14
CAAAA14053-22
1-Bromo-3,5-difluorobenzene 98%
1-Bromo-3,5-difluorobenzene
Purity: 98%
CAS No.: 461-96-1
Molecular Formula: C6H3BrF2
Formula Weight: 192.99
Flash Point: 44°(111°F)
Refractive Index: 1.4990
Melting Point: -27°
Boiling Point: 140-141°
Beilstein Reference: 1931520
UN No.: 1993
MDL No.: MFCD00010305
Formula:
C₆H₃BrF₂ Boiling Pt: 140…141 °C Melting Pt: –27 °C Density: 1.676 Flash Pt: 44 °C (111 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010305 CAS Number: 461-96-1 EINECS: 416-710-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...