(3-Bromopropyl)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-3-phenylpropane
A14050-0E
A14050-22
A14050-36
CAAAA14050-0EEA
1207.12
CAD
CAAAA14050-0E
CAAAA14050-22
CAAAA14050-36
(3-Bromopropyl)benzene 98%
(3-Bromopropyl)benzene
Purity: 98%
CAS No.: 637-59-2
Molecular Formula: C6H5CH2CH2CH2Br
Formula Weight: 199.09
Flash Point: 101°(213°F)
Refractive Index: 1.5460
Boiling Point: 219-220°
Beilstein Reference: 2205527
MDL No.: MFCD00000257
Formula:
BrH₂CCH₂CH₂(C₆H₅) MW: 199.09 g/mol Boiling Pt: 219 Density: 1.308 Flash Pt: 101 °C (213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000257 CAS Number: 637-59-2 EINECS: 211-294-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...