2,4-Difluorophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14044-30
A14044-18
CAAAAA14044-30EA
741.02
CAD
CAAAAA14044-30
CAAAA14044-18
2,4-Difluorophenol 98+%
2,4-Difluorophenol
Purity: 98+%
CAS No.: 367-27-1
Molecular Formula: C6H4F2O
Formula Weight: 130.09
Flash Point: 56°(132°F)
Refractive Index: 1.4860
Odor: Phenol-like
Melting Point: 21-24°
Boiling Point: 52-54°/14mm
Beilstein Reference: 1934803
UN No.: 2921
MDL No.: MFCD00009715
Formula:
C₆H₄F₂O Boiling Pt: 52…54 °C/14 mm Melting Pt: 21…24 °C Density: 1.362 Flash Pt: 56 °C (132 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009715 CAS Number: 367-27-1 EINECS: 206-688-0 UN: 2925 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...