Adamantane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tricyclo[3.3.1.1(3,7)]decane

A14041-0B A14041-18 A14041-30
CAAAAA14041-0BEA 1113.9 CAD
CAAAAA14041-0B CAAAA14041-18 CAAAA14041-30
Adamantane 98%
Adamantane

Purity: 98% CAS No.: 281-23-2 Molecular Formula: C10H16 Formula Weight: 136.24 Melting Point: 266-268°(sealed tube) Beilstein Reference: 1901173 Merck Reference: 14,149 MDL No.: MFCD00074719

Formula: C₁₀H₁₆
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: 266…268 °C (sealed tube)
Density: 1.070
Storage Temperature: Ambient
MDL Number: MFCD00074719
CAS Number: 281-23-2
EINECS: 206-001-4
UN: 3077
ADR: 9,III
Merck Index: 14,149

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR