4-Methylquinoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lepidine
A14040-09
A14040-18
A14040-30
CAAAA14040-09EA
71.1
CAD
CAAAA14040-09
CAAAA14040-18
CAAAA14040-30
4-Methylquinoline 97%
4-Methylquinoline
Purity: 97%
CAS No.: 491-35-0
Molecular Formula: C10H9N
Formula Weight: 143.19
Flash Point:
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 260…263 °C Melting Pt: 9…10 °C Density: 1.083 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006784 CAS Number: 491-35-0 EINECS: 207-734-2 Merck Index: 14,5441 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...