1-Bromoheptane 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14031-0B
A14031-0I
A14031-30
CAAAA14031-0BEA
268.6
CAD
CAAAA14031-0B
CAAAA14031-0I
CAAAA14031-30
1-Bromoheptane 98+%
1-Bromoheptane
Purity: 98+%
CAS No.: 629-04-9
Molecular Formula: CH3(CH2)6Br
Formula Weight: 179.11
Flash Point: 60°(140°F)
Refractive Index: 1.4499
Odor: Recognizable
Melting Point: -58°
Boiling Point: 179-180°
Beilstein Reference: 1697160
UN No.: 1993
MDL No.: MFCD00000273
Formula:
BrH₂C(CH₂)₅CH₃ Boiling Pt: 179…180 °C Melting Pt: –58 °C Density: 1.139 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000273 CAS Number: 629-04-9 EINECS: 211-068-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...