1-Bromo-3-chlorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14026-22
A14026-36
CAAAA14026-22EA
121.66
CAD
CAAAA14026-22
CAAAA14026-36
1-Bromo-3-chlorobenzene 99%
1-Bromo-3-chlorobenzene
Purity: 99%
CAS No.: 108-37-2
Molecular Formula: C6H4BrCl
Formula Weight: 191.46
Flash Point: 80°(176°F)
Refractive Index: 1.5770
Odor: Aromatic
Melting Point: -22°
Boiling Point: 195-196°
Beilstein Reference: 1446353
MDL No.: MFCD00000568
Formula:
C₆H₄BrCl Boiling Pt: 195…196 °C Melting Pt: –22 °C Density: 1.630 Flash Pt: 80 °C(176 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000568 CAS Number: 108-37-2 EINECS: 203-575-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...