4-Isopropylbenzaldehyde tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cuminaldehyde

A14019-14 A14019-22 A14019-36
CAAAA14019-14EA 48.66 CAD
CAAAA14019-14 CAAAA14019-22 CAAAA14019-36
4-Isopropylbenzaldehyde tech. 90%, Technical Grade
4-Isopropylbenzaldehyde

Purity: 90% CAS No.: 122-03-2 Molecular Formula: C10H12O Formula Weight: 148.21 Flash Point: 93°(199°F) Refractive Index: 1.5300 Boiling Point: 234-235° Beilstein Reference: 636547 Merck Reference: 14,2621 MDL No.: MFCD00006953

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 235
Density: 0.979
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006953
CAS Number: 122-03-2
EINECS: 204-516-9
Merck Index: 14,2621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR