Chloroacetic anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00000929
Beilstein: 774533
Freely soluble in ether, chloroform. Slightly soluble in benzene. Insoluble in cold ligroin
Purity: 97%
CAS No.: 541-88-8
Molecular Formula: (ClCH2CO)2O
Formula Weight: 170.98
Flash Point: 143°(289°F)
Melting Point: 56-59°
Boiling Point: 110-112°/10mm
Solubility: Freely soluble in ether, chloroform. Slightly soluble in benzene. Insoluble in cold ligroin
Beilstein Reference: 774533
Merck Reference: 14,2113
UN No.: 3261
MDL No.: MFCD00000929
Formula:
C₄H₄Cl₂O₃ Boiling Pt: 110…112 °C/10 mm Melting Pt: 56…59 °C Density: 1.5497 g/cm³ (20 °C) Flash Pt: 143 °C (289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000929 CAS Number: 541-88-8 EINECS: 208-794-2 UN: 2923 ADR: 8,III Merck Index: 14,2113 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...