2,5-Difluorobenzonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14014-06 A14014-14
CAAAA14014-06EA 73.31 CAD
CAAAA14014-06 CAAAA14014-14
2,5-Difluorobenzonitrile 98+%
2,5-Difluorobenzonitrile

Purity: 98+% CAS No.: 64248-64-2 Molecular Formula: C7H3F2N Formula Weight: 139.11 Flash Point: 77°(170°F) Melting Point: 31-35° Boiling Point: 188° Beilstein Reference: 2085640 UN No.: 3439 MDL No.: MFCD00001777

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 188 °C
Melting Pt: 31…35 °C
Flash Pt: 77 °C (170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001777
CAS Number: 64248-64-2
EINECS: 264-753-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR