3,3-Dimethyl-1-butyne 98%

Supplier: Thermo Scientific Chemicals
Danger

A14013-06 A14013-14
CAAAA14013-06EA 142.52 CAD
CAAAA14013-06 CAAAA14013-14
3,3-Dimethyl-1-butyne 98%
3,3-Dimethyl-1-butyne

Purity: 98% CAS No.: 917-92-0 Molecular Formula: C6H10 Formula Weight: 82.15 Storage Temperature: 2°C to 8°C Flash Point: -34°(-29°F) Refractive Index: 1.3745 Odor: Recognizable Melting Point: -78° Boiling Point: 37-38° Beilstein Reference: 1697100 UN No.: 3295 MDL No.: MFCD00008852

Formula: C₆H₁₀
Boiling Pt: 37…38 °C
Melting Pt: –78 °C
Density: 0.671
Flash Pt: <–34 °C (–29 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008852
CAS Number: 917-92-0
EINECS: 213-035-3
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR