3-Picoline 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
β-Picoline, 3-Methylpyridine
A14012-0F
A14012-AE
CAAAA14012-0FEA
244.9
CAD
CAAAA14012-0F
CAAAA14012-AE
3-Picoline 99%
3-Picoline
Purity: 99%
CAS No.: 108-99-6
Molecular Formula: C6H7N
Formula Weight: 93.13
Flash Point: 36°(96°F)
Refractive Index: 1.5055
Melting Point: -18°
Boiling Point: 143-144°
Beilstein Reference: 1366
Merck Reference: 14,7401
UN No.: 2313
MDL No.: MFCD00006402
Formula:
C₆H₇N MW: 93.13 g/mol Boiling Pt: 143…144 °C Melting Pt: –18 °C Density: 0.955 Flash Pt: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006402 CAS Number: 108-99-6 EINECS: 203-636-9 UN: 2313 ADR: 3,III Merck Index: 14,7401 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...