10-Undecen-1-ol 99%
Supplier: Thermo Scientific Chemicals
Warning
|
A14002-14
A14002-22
A14002-36
CAAAA14002-14EA
68.89
CAD
CAAAA14002-14
CAAAA14002-22
CAAAA14002-36
10-Undecen-1-ol 99%
10-Undecen-1-ol
Purity: 99%
CAS No.: 112-43-6
Molecular Formula: C11H22O
Formula Weight: 170.30
Flash Point: 93°(199°F)
Refractive Index: 1.4500
Melting Point: -3°
Boiling Point: 132-133°/15mm
Beilstein Reference: 1753225
MDL No.: MFCD00004750
Formula:
C₁₁H₂₂O Boiling Pt: 132…133 °C/15 mm Melting Pt: –3 °C Density: 0.848 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004750 CAS Number: 112-43-6 EINECS: 203-971-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...