Disodium hydrogen citrate sesquihydrate 99%

Supplier: Thermo Scientific Chemicals

A14001-0B A14001-0I A14001-30
CAAAA14001-0BEA 169.06 CAD
CAAAA14001-0B CAAAA14001-0I CAAAA14001-30
Disodium hydrogen citrate sesquihydrate 99%
di-Sodium hydrogen citrate sesquihydrate

Purity: 99% CAS No.: 6132-05-4 Molecular Formula: C6H6Na2O7.1.5H2O Formula Weight: 263.11 (236.09anhy) Melting Point: 149° dec. MDL No.: MFCD00150445

Formula: C₆H₆Na₂O₇·1.5H₂O
Melting Pt: 149 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150445
CAS Number: 6132-05-4
EINECS: 205-623-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR