1,5-Pentanediol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Pentane-1,5-diol, 1,5-Dihydroxy pentane
A13998-0E
A13998-22
A13998-36
CAAAA13998-0EEA
451.88
CAD
CAAAA13998-0E
CAAAA13998-22
CAAAA13998-36
1,5-Pentanediol 97%
1,5-Pentanediol
Purity: 97%
CAS No.: 111-29-5
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 129°(264°F)
Refractive Index: 1.4500
Melting Point: -16°
Boiling Point: 241-242°
Beilstein Reference: 1560130
Merck Reference: 14,7117
MDL No.: MFCD00002978
Formula:
C₅H₁₂O₂ Boiling Pt: 241…242 °C Melting Pt: –16 °C Density: 0.992 Flash Pt: 129 °C(264 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002978 CAS Number: 111-29-5 EINECS: 203-854-4 Merck Index: 14,7117 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...