4-tert-Butylbenzoic acid 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13997-0B
A13997-0I
A13997-30
CAAAA13997-0BEA
164.32
CAD
CAAAA13997-0B
CAAAA13997-0I
CAAAA13997-30
4-tert-Butylbenzoic acid 99+%
4-tert-Butylbenzoic acid
Purity: 99+%
CAS No.: 98-73-7
Molecular Formula: C11H14O2
Formula Weight: 178.23
Flash Point: 180°(356°F)
Melting Point: 164-168°
Boiling Point: 280° dec.
Beilstein Reference: 607545
UN No.: 2811
MDL No.: MFCD00002563
Formula:
C₁₁H₁₄O₂ Boiling Pt: 280 °C (dec.) Melting Pt: 164…168 °C Density: 0.6 g/cm³ (20 °C) Flash Pt: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002563 CAS Number: 98-73-7 EINECS: 202-696-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...