4-Pentenoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Allylacetic acid, Pent-4-enoic acid
A13991-09
A13991-14
A13991-22
CAAAAA13991-09EA
71.73
CAD
CAAAAA13991-09
CAAAA13991-14
CAAAA13991-22
4-Pentenoic acid 98%
4-Pentenoic acid
MDL: MFCD00004408
Beilstein: 1633696
Soluble in alcohol, Slightly soluble in water
Purity: 98%
CAS No.: 591-80-0
Molecular Formula: H2C=CH(CH2)2CO2H
Formula Weight: 100.12
Flash Point: 89°(192°F)
Refractive Index: 1.4283
Odor: Pungent
Melting Point: -22°
Boiling Point: 187-189°
Solubility: Soluble in alcohol, Slightly soluble in water
Beilstein Reference: 1633696
UN No.: 3265
MDL No.: MFCD00004408
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 187…189 °C Melting Pt: –22 °C Density: 0.891 Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004408 CAS Number: 591-80-0 EINECS: 209-732-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...