3-Bromotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

A13990-14 A13990-22 A13990-36
CAAAA13990-14EA 87.22 CAD
CAAAA13990-14 CAAAA13990-22 CAAAA13990-36
3-Bromotoluene 98%
3-Bromotoluene

Purity: 98% CAS No.: 591-17-3 Molecular Formula: C7H7Br Formula Weight: 171.04 Flash Point: 75°(167°F) Refractive Index: 1.5520 Melting Point: -40° Boiling Point: 183-184° Beilstein Reference: 1903633 Merck Reference: 14,1439 MDL No.: MFCD00000085

Formula: C₇H₇Br
Boiling Pt: 183…184 °C
Melting Pt: –40 °C
Density: 1.410
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000085
CAS Number: 591-17-3
EINECS: 209-702-3
UN: 1993
ADR: 3,III
Merck Index: 14,1439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR