(Ethylthio)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl phenyl sulphide, Ethyl phenyl sulfide
A13987-22
A13987-06
A13987-14
CAAAAA13987-22EA
535.62
CAD
CAAAAA13987-22
CAAAA13987-06
CAAAA13987-14
(Ethylthio)benzene 98%
(Ethylthio)benzene
Purity: 98%
CAS No.: 622-38-8
Molecular Formula: C6H5SCH2CH3
Formula Weight: 138.23
Flash Point: 73°(163°F)
Refractive Index: 1.5655
Odor: Stench
Boiling Point: 204-205°
Beilstein Reference: 1905731
MDL No.: MFCD00009265
Formula:
C₈H₁₀S Boiling Pt: 204…205 °C Melting Pt: 206.33 °C Density: 1.020 Flash Pt: 73 °C(163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009265 CAS Number: 622-38-8 EINECS: 210-731-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...