Diphenylacetylene 99%

Supplier: Thermo Scientific Chemicals

A13979-06 A13979-14 A13979-22
CAAAA13979-06EA 55.62 CAD
CAAAA13979-06 CAAAA13979-14 CAAAA13979-22
Diphenylacetylene 99%
Diphenylacetylene

Purity: 99% CAS No.: 501-65-5 Molecular Formula: C14H10 Formula Weight: 178.23 Storage Temperature: 2°C to 8°C Odor: Odorless Boiling Point: 170°/19mm Beilstein Reference: 606478 Merck Reference: 14,9504 MDL No.: MFCD00004786

Formula: C₁₄H₁₀
Boiling Pt: 170 °C/19 mm
Melting Pt: 58…62 °C
Density: 0.990
Storage Temperature: Refrigerator
MDL Number: MFCD00004786
CAS Number: 501-65-5
EINECS: 207-926-6
Merck Index: 14,9504

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR