4-Fluoro-o-phenylenediamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,4-Diamino-1-fluorobenzene, 4-Fluorobenzene-1,2-diamine, 1,2-Diamino-4-fluorobenzene, 4-Fluoro-1,2-phenylenediamine
A13977-06
A13977-14
CAAAAA13977-22EA
3859.96
CAD
CAAAAA13977-22
CAAAA13977-06
CAAAA13977-14
4-Fluoro-o-phenylenediamine 97%
4-Fluoro-o-phenylenediamine
Purity: 97%
CAS No.: 367-31-7
Molecular Formula: C6H7FN2
Formula Weight: 126.13
Melting Point: 91-95°
Beilstein Reference: 2081075
UN No.: 2811
MDL No.: MFCD00042228
Formula:
C₆H₇FN₂ MW: 126.13 g/mol Boiling Pt: 250 °C (1013 hPa) Melting Pt: 91…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042228 CAS Number: 367-31-7 EINECS: 206-691-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...