Methyl pyruvate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13966-36
A13966-14
A13966-22
CAAAAA13966-36EA
570.38
CAD
CAAAAA13966-36
CAAAA13966-14
CAAAA13966-22
Methyl pyruvate 98%
Methyl pyruvate
Purity: 98%
CAS No.: 600-22-6
Molecular Formula: C4H6O3
Formula Weight: 102.09
Storage Temperature: 2°C to 8°C
Flash Point: 39°(102°F)
Refractive Index: 1.4040
Odor: Sweet
Boiling Point: 135-137°
Beilstein Reference: 1361953
Merck Reference: 14,8021
UN No.: 3272
MDL No.: MFCD00008754
Formula:
C₄H₆O₃ Boiling Pt: 135…137 °C Density: 1.123 Flash Pt: 39 °C (102 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008754 CAS Number: 600-22-6 EINECS: 209-987-4 UN: 3272 ADR: 3,III Merck Index: 14,8021 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...