2-Amino-4-nitrophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-5-nitroaniline

A13964-14 A13964-22
CAAAAA13964-36EA 925.69 CAD
CAAAAA13964-36 CAAAA13964-14 CAAAA13964-22
2-Amino-4-nitrophenol 98%
2-Amino-4-nitrophenol

Purity: 98% CAS No.: 99-57-0 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: 141-145° Beilstein Reference: 776533

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 141…145 °C
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007695
CAS Number: 99-57-0
EINECS: 202-767-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR