4'-Fluoroacetophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13950-14
A13950-22
A13950-36
CAAAA13950-14EA
51.82
CAD
CAAAA13950-14
CAAAA13950-22
CAAAA13950-36
4'-Fluoroacetophenone 99%
4'-Fluoroacetophenone
Purity: 99%
CAS No.: 403-42-9
Molecular Formula: C8H7FO
Formula Weight: 138.14
Flash Point: 71°(159°F)
Refractive Index: 1.5110
Melting Point: 4°
Boiling Point: 194-196°
Beilstein Reference: 386013
MDL No.: MFCD00000354
Formula:
C₈H₇FO Boiling Pt: 194…196 °C Melting Pt: 4 °C Density: 1.141 Flash Pt: 71 °C(159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000354 CAS Number: 403-42-9 EINECS: 206-960-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...