5-Fluoro-2-nitrotoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Fluoro-3-methyl-4-nitrobenzene
A13944-09
A13944-18
CAAAA13944-09EA
60.83
CAD
CAAAA13944-09
CAAAA13944-18
5-Fluoro-2-nitrotoluene 98+%
5-Fluoro-2-nitrotoluene
Purity: 98+%
CAS No.: 446-33-3
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Flash Point: 87°(188°F)
Refractive Index: 1.5280
Melting Point: ca 20°
Boiling Point: 97-98°/10mm
Beilstein Reference: 2046652
UN No.: 2810
MDL No.: MFCD00007283
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 97…98 °C/10 mm Melting Pt: ∼ 20 °C Density: 1.270 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007283 CAS Number: 446-33-3 EINECS: 207-164-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...