α,α'-Dibromo-p-xylene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Xylylene dibromide, 1,4-Bis(bromomethyl)benzene

A13939-09 A13939-18
CAAAA13939-09EA 85.64 CAD
CAAAA13939-09 CAAAA13939-18
α,α'-Dibromo-p-xylene 97%
α,α'-Dibromo-p-xylene

Purity: 97% CAS No.: 623-24-5 Molecular Formula: C8H8Br2 Formula Weight: 263.96 Melting Point: 144-148° Boiling Point: 245° UN No.: 3261 MDL No.: MFCD00000182

Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 245 °C
Melting Pt: 144…148 °C
Density: 2.012
Storage Temperature: Ambient
MDL Number: MFCD00000182
CAS Number: 623-24-5
EINECS: 210-781-1
UN: 2928
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR