Isopropyl-4-hydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

A13930-09 A13930-18 A13930-30
CAAAA13930-09EA 89.9 CAD
CAAAA13930-09 CAAAA13930-18 CAAAA13930-30
Isopropyl-4-hydroxybenzoate 98%
Isopropyl 4-hydroxybenzoate

Purity: 98% CAS No.: 4191-73-5 Molecular Formula: C10H12O3 Formula Weight: 180.21 Melting Point: 84-86° Beilstein Reference: 2641564

Formula: C₁₀H₁₂O₃
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00016468
CAS Number: 4191-73-5
EINECS: 224-069-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR