(2-Hydroxyphenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxyphenylacetic acid

A13924-09 A13924-18
CAAAA13924-09EA 75.37 CAD
CAAAA13924-09 CAAAA13924-18
(2-Hydroxyphenyl)acetic acid 98%
(2-Hydroxyphenyl)acetic acid

Purity: 98% CAS No.: 614-75-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 145-147° Beilstein Reference: 908000

Formula: (C₆H₄OH)CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 328
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00004323
CAS Number: 614-75-5
EINECS: 210-393-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR