1,10-Decanediol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,10-Dihydroxy decane
A13923-22
A13923-36
CAAAA13923-22EA
87.53
CAD
CAAAA13923-22
CAAAA13923-36
1,10-Decanediol 97%
1,10-Decanediol
Purity: 97%
CAS No.: 112-47-0
Molecular Formula: HO(CH2)10OH
Formula Weight: 174.28
Flash Point: 152°(306°F)
Melting Point: 71-74°
Boiling Point: 297°
Beilstein Reference: 1698975
Merck Reference: 14,2849
MDL No.: MFCD00004749
Formula:
C₁₀H₂₂O₂ MW: 174.28 g/mol Boiling Pt: 297 Melting Pt: 72 Density: 1.080 Flash Pt: 152 °C (306 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004749 CAS Number: 112-47-0 EINECS: 203-975-2 Merck Index: 14,2849 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...