3,5-Dichloroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13922-18
A13922-30
CAAAA13922-18EA
73.15
CAD
CAAAA13922-18
CAAAA13922-30
3,5-Dichloroaniline 98%
3,5-Dichloroaniline
Purity: 98%
CAS No.: 626-43-7
Molecular Formula: C6H5Cl2N
Formula Weight: 162.02
Flash Point: 133°(271°F)
Melting Point: 49-53°
Boiling Point: 259-260°/741mm
Beilstein Reference: 636492
UN No.: 3442
MDL No.: MFCD00007774
Formula:
C₆H₅Cl₂N MW: 162.02 g/mol Boiling Pt: 259…260 °C/741 mm Melting Pt: 49…53 °C Density: 1.58 Flash Pt: 133 °C (271 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007774 CAS Number: 626-43-7 EINECS: 210-948-9 UN: 3442 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...