2-Hydroxy-5-nitropyridine 98+%

Supplier: Thermo Scientific Chemicals
Warning

A13921-14 A13921-22
CAAAA13921-14EA 84.21 CAD
CAAAA13921-14 CAAAA13921-22
2-Hydroxy-5-nitropyridine 98+%
2-Hydroxy-5-nitropyridine

Purity: 98+% CAS No.: 5418-51-9 Molecular Formula: C5H4N2O3 Formula Weight: 140.10 Melting Point: 188-191° Beilstein Reference: 120352

Formula: C₅H₄N₂O₃
Boiling Pt: 180…200 °C (0,2 mmHg)
Melting Pt: 188…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00006276
CAS Number: 5418-51-9
EINECS: 226-525-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR