Diphenyl diselenide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13919-06
A13919-14
A13919-22
CAAAA13919-06EA
92.27
CAD
CAAAA13919-06
CAAAA13919-14
CAAAA13919-22
Diphenyl diselenide 98%
Diphenyl diselenide
Purity: 98%
CAS No.: 1666-13-3
Molecular Formula: C12H10Se2
Formula Weight: 312.13
Melting Point: 61-63°
Beilstein Reference: 2047179
UN No.: 3283
MDL No.: MFCD00003001
Formula:
C₁₂H₁₀Se₂ Boiling Pt: 377 °C (1013 hPa) Melting Pt: 61…63 °C Density: 1.32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003001 CAS Number: 1666-13-3 EINECS: 216-780-2 UN: 3283 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...