3-Pyridone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13910-22
A13910-36
A13910-18
CAAAAA13910-22EA
86.27
CAD
CAAAAA13910-22
CAAAAA13910-36
CAAAA13910-0B
CAAAA13910-18
3-Pyridone 98%
3-Pyridone
Purity: 98%
CAS No.: 109-00-2
Molecular Formula: C5H5NO
Formula Weight: 95.10
Flash Point: 173°(343°F)
Melting Point: 125-128°
Boiling Point: 180°/22mm
Beilstein Reference: 105699
MDL No.: MFCD00006378
Formula:
C₅H₅NO Boiling Pt: 180 °C/22 mm Melting Pt: 125…128 °C Density: 1.2 g/cm³ (-70 °C) Flash Pt: 173 °C (343 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006378 CAS Number: 109-00-2 EINECS: 203-637-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...