3-Ethoxybenzonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13908-06
A13908-14
CAAAA13908-06EA
47.4
CAD
CAAAA13908-06
CAAAA13908-14
3-Ethoxybenzonitrile 95%
3-Ethoxybenzonitrile
Purity: 95%
CAS No.: 25117-75-3
Molecular Formula: C9H9NO
Formula Weight: 147.18
Flash Point:
Formula:
C₉H₉NO Boiling Pt: 241…244 °C Density: 1.047 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001805 CAS Number: 25117-75-3 EINECS: 246-632-2 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...