(R)-(-)-Carvone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, L-p-Mentha-1(6),8-dien-2-one, (R)-(-)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one, L(-)-Carvone
A13900-0B
A13900-18
A13900-30
CAAAA13900-0BEA
652.54
CAD
CAAAA13900-0B
CAAAA13900-18
CAAAA13900-30
(R)-(-)-Carvone 98%
(R)-(-)-Carvone
Purity: 98%
CAS No.: 6485-40-1
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 93°(199°F)
Physical Form: Liquid
Density: 0.959
Refractive Index: 1.4990
Appearance: Colorless
Odor: Pleasant
Boiling Point: 228-230°
Beilstein Reference: 2206714
Optical Rotation: -61° (neat)
MDL No.: MFCD00001578
Formula:
C₁₀H₁₄O MW: 150.22 g/mol Boiling Pt: 228…230 °C Melting Pt: 25,2 °C Density: 0.959 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001578 CAS Number: 6485-40-1 EINECS: 229-352-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...