2-Methylanisole 99%

Supplier: Thermo Scientific Chemicals
Warning

A13897-18 A13897-30
CAAAA13897-0BEA 1587.52 CAD
CAAAA13897-0B CAAAA13897-18 CAAAA13897-30
2-Methylanisole 99%
2-Methylanisole

Purity: 99% CAS No.: 578-58-5 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 51°(123°F) Refractive Index: 1.5170 Boiling Point: 170-172° Beilstein Reference: 1857415 UN No.: 3271 MDL No.: MFCD00008373

Formula: C₈H₁₀O
Boiling Pt: 170…172 °C
Density: 0.985
Flash Pt: 51 °C (123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008373
CAS Number: 578-58-5
EINECS: 209-426-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR