trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2E)-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, (E)-Ferulic acid, Trans-3-(4-hydroxy-3-methoxyphenyl)acrylic acid, trans-Ferulic acid

A13890-14 A13890-22 A13890-36
CAAAA13890-14EA 111.55 CAD
CAAAA13890-14 CAAAA13890-22 CAAAA13890-36
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid (trans-) 99%
trans-3-(4-Hydroxy-3-methoxyphenyl)acrylic acid

A chemopreventive analogue of caffeic acid


Purity: 99% CAS No.: 537-98-4 Molecular Formula: C10H10O4 Formula Weight: 194.19 Melting Point: 170-175° Beilstein Reference: 1371483 Merck Reference: 14,4062 MDL No.: MFCD00004400

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00004400
CAS Number: 537-98-4
EINECS: 208-679-7
Merck Index: 14,4062

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR