1,2-Dichlorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13881-0C
A13881-0E
A13881-36
CAAAA13881-0CEA
837.4
CAD
CAAAA13881-0C
CAAAA13881-0E
CAAAA13881-36
1,2-Dichlorobenzene 99%
1,2-Dichlorobenzene
Purity: 99%
CAS No.: 95-50-1
Molecular Formula: C6H4Cl2
Formula Weight: 147.00
Flash Point: 68°(154°F)
Refractive Index: 1.5510
Odor: Pungent
Melting Point: -18°
Boiling Point: 180°
Beilstein Reference: 606078
Merck Reference: 14,3056
UN No.: 1591
MDL No.: MFCD00000535
Formula:
C₆H₄Cl₂ Boiling Pt: 180 °C Melting Pt: –18 °C Density: 1.306 Flash Pt: 68 °C(154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000535 CAS Number: 95-50-1 EINECS: 202-425-9 UN: 1591 ADR: "6.1",III Merck Index: 14,3056 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...