5-Chloro-4-fluoro-2-nitroaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Amino-3-chlor-4-fluor-6-nitrobenzene
A13879-14
A13879-06
CAAAAA13879-14EA
532.46
CAD
CAAAAA13879-14
CAAAA13879-03
CAAAA13879-06
5-Chloro-4-fluoro-2-nitroaniline 97%
5-Chloro-4-fluoro-2-nitroaniline
Purity: 97%
CAS No.: 104222-34-6
Molecular Formula: C6H4ClFN2O2
Formula Weight: 190.56
Melting Point: 141-145°
Beilstein Reference: 7202993
UN No.: 2811
MDL No.: MFCD00052698
Formula:
C₆H₄ClFN₂O₂ MW: 190.56 g/mol Melting Pt: 141…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052698 CAS Number: 104222-34-6 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...