4'-Nitroacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
A13877-18
A13877-30
CAAAA13877-0BEA
569.38
CAD
CAAAA13877-0B
CAAAA13877-18
CAAAA13877-30
4'-Nitroacetophenone 98%
4'-Nitroacetophenone
Purity: 98%
CAS No.: 100-19-6
Molecular Formula: C8H7NO3
Formula Weight: 165.15
Flash Point: 149°(300°F)
Physical Form: Cryst powder
Odor: Characteristic
Melting Point: 77-81°
Boiling Point: 201-202°
Beilstein Reference: 607777
MDL No.: MFCD00007355
Formula:
C₈H₇NO₃ MW: 165.15 g/mol Boiling Pt: 201…202 °C Melting Pt: 77…81 °C Flash Pt: 149 °C (300 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007355 CAS Number: 100-19-6 EINECS: 202-827-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...