(±)-2-Methylvaleric acid 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(±)-2-Methylpentanoic acid, 2-Methylpentanoic Acid, 2-Methylvaleric Acid
A13873-0E
A13873-22
CAAAA13873-0EEA
802.64
CAD
CAAAA13873-0E
CAAAA13873-22
(±)-2-Methylvaleric acid 98+%
(±)-2-Methylvaleric acid
Purity: 98+%
CAS No.: 97-61-0
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 91°(195°F)
Refractive Index: 1.4140
Odor: Unpleasant
Melting Point: -85°
Boiling Point: 196-197°
Beilstein Reference: 1720655
UN No.: 3265
MDL No.: MFCD00002671
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 196…197 °C Melting Pt: –85 °C Density: 0.930 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002671 CAS Number: 97-61-0 EINECS: 202-594-9 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...